ENA341296044 Enamine

1-methyl-3-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06805246
  • Cas Number : 177409-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 170-172
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92745
Availability details for
Quantity
  • Availability
  • Add to Cart