ENA341296049 Enamine

5-(furan-2-yl)-1,1-dioxo-2H-1lambda6,2,6-thiadiazine-3-carboxylic acid

  • MDL number MFCD09473397
  • Cas Number : 1011397-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92750
Availability details for
Quantity
  • Availability
  • Add to Cart