ENA341296050 Enamine

4-[(4-chloro-1H-pyrazol-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06803412
  • Cas Number : 1006485-93-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92751

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92751
Availability details for
Quantity
  • Availability
  • Add to Cart