ENA341296054 Enamine

1-[(4-chlorophenyl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06805382
  • Cas Number : 1006452-71-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92755
Availability details for
Quantity
  • Availability
  • Add to Cart