ENA341296055 Enamine

1-ethyl-7-methyl-4-oxo-2-sulfanyl-1H,4H-pyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08559036
  • Cas Number : 938002-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92756

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92756
Availability details for
Quantity
  • Availability
  • Add to Cart