ENA341296057 Enamine

3-[(4-fluorophenoxy)methyl]benzoic acid

  • MDL number MFCD03421920
  • Cas Number : 869950-21-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92758

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92758
Availability details for
Quantity
  • Availability
  • Add to Cart