ENA341296060 Enamine

2-{[(1-ethyl-1H-pyrazol-4-yl)methyl]amino}benzoic acid

  • MDL number MFCD08558988
  • Cas Number : 1006348-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92761
Availability details for
Quantity
  • Availability
  • Add to Cart