ENA341296073 Enamine

4-[(3-amino-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD04969600
  • Cas Number : 1006348-84-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92774

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92774
Availability details for
Quantity
  • Availability
  • Add to Cart