ENA341296077 Enamine

3-(2-chloro-6-fluorophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD05995808
  • Cas Number : 712347-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92778

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92778
Availability details for
Quantity
  • Availability
  • Add to Cart