ENA341296084 Enamine

2-(3-cyclopropyl-1H-pyrazol-1-yl)propanoic acid

  • MDL number MFCD04970949
  • Cas Number : 1006446-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92785
Availability details for
Quantity
  • Availability
  • Add to Cart