ENA341296085 Enamine

5-(3,4-dimethylphenoxymethyl)thiophene-2-carboxylic acid

  • MDL number MFCD06804701
  • Cas Number : 934080-82-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92786

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92786
Availability details for
Quantity
  • Availability
  • Add to Cart