ENA341296086 Enamine

7-cyclopropylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04053086
  • Cas Number : 832114-69-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92787
Availability details for
Quantity
  • Availability
  • Add to Cart