ENA341296087 Enamine

2-{3-methyl-5-nitro-1H-pyrazolo[3,4-b]pyridin-1-yl}acetic acid

  • MDL number MFCD09473502
  • Cas Number : 1011398-78-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92788

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92788
Availability details for
Quantity
  • Availability
  • Add to Cart