ENA341296089 Enamine

3-(1-methyl-1H-pyrazol-3-yl)propanoic acid

  • MDL number MFCD06808504
  • Cas Number : 1006440-24-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92790

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92790
Availability details for
Quantity
  • Availability
  • Add to Cart