ENA341296091 Enamine

2-{[4-cyclopropyl-6-(trifluoromethyl)pyrimidin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD03422182
  • Cas Number : 861434-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92792

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92792
Availability details for
Quantity
  • Availability
  • Add to Cart