ENA341296093 Enamine

5-chloro-6-cyclopropyl-1,3-dimethyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD09701519
  • Cas Number : 1018142-01-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92794

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92794
Availability details for
Quantity
  • Availability
  • Add to Cart