ENA341296094 Enamine

4-[(4-chloro-1H-pyrazol-1-yl)methoxy]benzoic acid

  • MDL number MFCD04967875
  • Cas Number : 1006474-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92795

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92795
Availability details for
Quantity
  • Availability
  • Add to Cart