ENA341296100 Enamine

3-[(3-amino-5-methyl-1H-pyrazol-1-yl)methyl]benzoic acid

  • MDL number MFCD04969607
  • Cas Number : 1006433-75-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92801

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92801
Availability details for
Quantity
  • Availability
  • Add to Cart