ENA341296101 Enamine

1-butyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD06805217
  • Cas Number : 1006493-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92802

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92802
Availability details for
Quantity
  • Availability
  • Add to Cart