ENA341296102 Enamine

2-[3-(1,3-dimethyl-1H-pyrazol-4-yl)-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD04970841
  • Cas Number : 1006464-37-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92803
Availability details for
Quantity
  • Availability
  • Add to Cart