ENA341296107 Enamine

2-{[4-(1-methyl-1H-pyrazol-4-yl)pyrimidin-2-yl]sulfanyl}acetic acid

  • MDL number MFCD04970322
  • Cas Number : 1006334-23-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92808

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92808
Availability details for
Quantity
  • Availability
  • Add to Cart