ENA341296108 Enamine

3-(1-ethyl-3-methyl-1H-pyrazol-4-yl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD06739306
  • Cas Number : 907990-49-2
  • Molecular Weight : 223.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92809
Availability details for
Quantity
  • Availability
  • Add to Cart