ENA341296110 Enamine

3-{[(1,3-dimethyl-1H-pyrazol-4-yl)methyl]sulfanyl}propanoic acid

  • MDL number MFCD04970141
  • Cas Number : 1006352-76-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92811
Availability details for
Quantity
  • Availability
  • Add to Cart