ENA341296113 Enamine

7-phenylpyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD06739327
  • Cas Number : 886503-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92814
Availability details for
Quantity
  • Availability
  • Add to Cart