ENA341296121 Enamine

4-[(1H-pyrazol-1-yl)methyl]thiophene-2-carboxylic acid

  • MDL number MFCD06803411
  • Cas Number : 1006334-13-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92822

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92822
Availability details for
Quantity
  • Availability
  • Add to Cart