ENA341296124 Enamine

3-(4-methoxyphenyl)-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD04124288
  • Cas Number : 618383-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92825

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92825
Availability details for
Quantity
  • Availability
  • Add to Cart