ENA341296127 Enamine

2-[(5-cyclohexyl-4-methyl-4H-1,2,4-triazol-3-yl)sulfanyl]acetic acid

  • MDL number MFCD02588700
  • Cas Number : 462066-86-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92828

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92828
Availability details for
Quantity
  • Availability
  • Add to Cart