ENA341296128 Enamine

3-[(4-chloro-1H-pyrazol-1-yl)methyl]-4-methoxybenzoic acid

  • MDL number MFCD06653196
  • Cas Number : 956786-68-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92829

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92829
Availability details for
Quantity
  • Availability
  • Add to Cart