ENA341296135 Enamine

3-(diethylcarbamoyl)-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06739384
  • Cas Number : 1004727-28-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92836

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92836
Availability details for
Quantity
  • Availability
  • Add to Cart