ENA341296139 Enamine

4-[(3-fluorophenoxy)methyl]-5-methyl-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD06805574
  • Cas Number : 934063-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92840
Availability details for
Quantity
  • Availability
  • Add to Cart