ENA341296141 Enamine

7-methyl-2,4-dioxo-1H,2H,3H,4H-pyrido[2,3-d]pyrimidine-5-carboxylic acid

  • MDL number MFCD08558295
  • Cas Number : 116081-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92842
Availability details for
Quantity
  • Availability
  • Add to Cart