ENA341296146 Enamine

7-(trifluoromethyl)-4H,5H,6H,7H-pyrazolo[1,5-a]pyrimidine-2-carboxylic acid

  • MDL number MFCD04969254
  • Cas Number : 869948-09-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92847
Availability details for
Quantity
  • Availability
  • Add to Cart