ENA341296147 Enamine

2-[3,5-bis(difluoromethyl)-4-nitro-1H-pyrazol-1-yl]acetic acid

  • MDL number MFCD09468201
  • Cas Number : 1171731-28-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92848
Availability details for
Quantity
  • Availability
  • Add to Cart