ENA341296153 Enamine

4-(4-bromo-3,5-dimethyl-1H-pyrazol-1-yl)butanoic acid

  • MDL number MFCD06167690
  • Cas Number : 898050-16-3
  • Molecular Weight : 261.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92854
Availability details for
Quantity
  • Availability
  • Add to Cart