ENA341296154 Enamine

3-(3-bromophenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD05995865
  • Cas Number : 712347-61-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92855

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92855
Availability details for
Quantity
  • Availability
  • Add to Cart