ENA341296156 Enamine

3-(4-methoxyphenyl)-4,5-dihydro-1,2-oxazole-5-carboxylic acid

  • MDL number MFCD06011104
  • Cas Number : 473253-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. Normal

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92857
Availability details for
Quantity
  • Availability
  • Add to Cart