ENA341296161 Enamine

5-[(3-amino-1H-pyrazol-1-yl)methyl]furan-2-carboxylic acid

  • MDL number MFCD04969604
  • Cas Number : 1006348-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92862

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92862
Availability details for
Quantity
  • Availability
  • Add to Cart