ENA341296162 Enamine

5-methyl-4-[(4-nitro-1H-pyrazol-1-yl)methyl]-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD06653271
  • Cas Number : 956758-72-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92863

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92863
Availability details for
Quantity
  • Availability
  • Add to Cart