ENA341296165 Enamine

1-(carboxymethyl)-3-nitro-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD06739328
  • Cas Number : 956439-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92866

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92866
Availability details for
Quantity
  • Availability
  • Add to Cart