ENA341296167 Enamine

1-(difluoromethyl)-3-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD04970129
  • Cas Number : 1006319-91-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92868
Availability details for
Quantity
  • Availability
  • Add to Cart