ENA341296173 Enamine

1-{1-phenyl-1H-pyrazolo[3,4-d]pyrimidin-4-yl}piperidine-4-carboxylic acid

  • MDL number MFCD08558311
  • Cas Number : 932162-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-92874

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92874
Availability details for
Quantity
  • Availability
  • Add to Cart