ENA341296176 Enamine

5-(4-chlorophenyl)-4-ethylthiophene-2-carboxylic acid

  • MDL number MFCD05861721
  • Cas Number : 861226-90-6
  • Molecular Weight : 266.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-92877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92877
Availability details for
Quantity
  • Availability
  • Add to Cart