ENA341296178 Enamine

2-[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]propanoic acid

  • MDL number MFCD04970962
  • Cas Number : 869942-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92879

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92879
Availability details for
Quantity
  • Availability
  • Add to Cart