ENA341296186 Enamine

4-amino-1,5-dimethyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08556300
  • Cas Number : 1006336-98-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92887
Availability details for
Quantity
  • Availability
  • Add to Cart