ENA341296192 Enamine

6-cyclopropyl-1-propyl-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08558139
  • Cas Number : 937597-47-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92893

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92893
Availability details for
Quantity
  • Availability
  • Add to Cart