ENA341296198 Enamine

2-[3-(trifluoromethyl)-1H,4H,5H,6H-cyclopenta[c]pyrazol-1-yl]acetic acid

  • MDL number MFCD02732817
  • Cas Number : 333309-24-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92899

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92899
Availability details for
Quantity
  • Availability
  • Add to Cart