ENA341296204 Enamine

3,4-dihydro-1H-2-benzothiopyran-1-carboxylic acid

  • MDL number MFCD01850935
  • Cas Number : 93689-97-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92906

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92906
Availability details for
Quantity
  • Availability
  • Add to Cart