ENA341296205 Enamine

1,2$l^{6}-oxathiolane-2,2,5-trione

  • MDL number MFCD07783392
  • Cas Number : 5961-88-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 69...71
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92907

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92907
Availability details for
Quantity
  • Availability
  • Add to Cart