ENA341296209 Enamine

2-fluoro-5-(furan-2-yl)aniline

  • MDL number MFCD14623706
  • Cas Number : 1248081-28-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 40...42
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-92911

Safety Information

msds https://www.enaminestore.com/catalog/EN300-92911
Availability details for
Quantity
  • Availability
  • Add to Cart