ENA341296210 Enamine

5-carbamoyl-1-methyl-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD12548031
  • Cas Number : 1174878-96-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-92425

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-92425
Availability details for
Quantity
  • Availability
  • Add to Cart